Yttrium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.127 mm

Supplier: BeanTown Chemical

211505-25X25MM 211505-50X50MM
BT211505-25X25EA 100 USD
BT211505-25X25 BT211505-50X50
Yttrium ≥99.9% (trace rare earth metals basis), foil, Thickness 0.127 mm
Yttrium

CAS: 7440-65-5; EC No: 231-174-8; MDL No: MFCD00011468 Solid; Molecular Formula: Y; MW: 88.91 Melting Point: 1522°; Boiling Point: 3338° Density (g/mL): 4.469


  • Solid

Formula: Y
MW: 88.91
Boiling Pt: 2927
Melting Pt: 1525
Density: 4.469 g/mL
MDL Number: MFCD00011468
CAS Number: 7440-65-5
EINECS: 231-174-8
Merck Index: 12,10240

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR