2-Indanone ≥98%
Supplier: BeanTown Chemical
Synonyms:
beta-Hydrindone, 2-Oxoindane, 1H-Inden-2(3H)-one
211485-100G
211485-25G
211485-5G
BT211485-100GEA
664
USD
BT211485-100G
BT211485-25G
BT211485-5G
2-Indanone ≥98%
2-Indanone
CAS: 615-13-4; EC No: 210-410-3; MDL No: MFCD00003792; RTECS: NK7535500
Crystalline; Molecular Formula: C9H8O ; MW: 132.16
Melting Point: 51-54°; Flash point: 100°C (212°F)
Hygroscopic
- RTECS: NK7535500
- Hygroscopic
- Crystalline
Formula:
C₉H₈O Melting Pt: 51…54 °C Flash Pt: 100 °C (212 °F) |
MDL Number:
MFCD00003792 CAS Number: 615-13-4 EINECS: 210-410-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...