Aluminium sulfate, anhydrous ≥99.99%
Supplier: BeanTown Chemical
Synonyms:
Aluminum sulphate, Aluminum sulfate
211470-100G
211470-25G
211470-5G
BT211470-100GEA
418.72
USD
BT211470-100G
BT211470-25G
BT211470-5G
Aluminium sulfate, anhydrous ≥99.99%
Aluminium sulphate
CAS: 10043-01-3; EC No: 233-135-0; MDL No: MFCD00003423; RTECS: BD1700000
Powder; Linear Formula: Al2(SO4)3 ; MW: 342.15
Melting Point: 770°
Density (g/mL): 2.71
Hygroscopic
- RTECS: BD1700000
- Hygroscopic
- Powder
Formula:
Al₂O₁₂S₃ Melting Pt: 770 °C Density: 2.71 g/mL |
MDL Number:
MFCD00003423 CAS Number: 10043-01-3 EINECS: 233-135-0 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...