N,N-Dimethyloctylamine ≥95%

Supplier: BeanTown Chemical

Synonyms: N,N-Dimethyloctan-1-amine

211390-10G 211390-250G 211390-50G
BT211390-10GEA 37.55 USD
BT211390-10G BT211390-250G BT211390-50G
N,N-Dimethyloctylamine ≥95%
N,N-Dimethyloctylamine

CAS: 7378-99-6; EC No: 230-939-3; MDL No: MFCD00009558 UN No: UN2922; Haz Class: 8 (6.1); Packing Group: III Liquid; Linear Formula: CH3(CH2)7N(CH3)2; Molecular Formula: C10H23N; MW: 157.30 Melting Point: -57°; Boiling Point: 195°; Flash point: 64°C (147°F) Density (g/mL): 0.765; Refractive Index: 1.424 Air Sensitive


  • Refractive index: 1.424
  • Air Sensitive
  • Liquid

Formula: C₁₀H₂₃N
MW: 157.3 g/mol
Boiling Pt: 195
Melting Pt: -57
Density: 0.765 g/mL
Flash Pt: 64 °C (147 °F)
MDL Number: MFCD00009558
CAS Number: 7378-99-6
EINECS: 230-939-3
UN: 2922
ADR: 8 III

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR