Titanium(IV) oxide ≥99.995% (mixture of rutile and anatase)

Supplier: BeanTown Chemical

Synonyms: Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide

211360-10G 211360-250G 211360-50G
BT211360-10GEA 79 USD
BT211360-10G BT211360-250G BT211360-50G
Titanium(IV) oxide ≥99.995% (mixture of rutile and anatase)
Titanium (IV) oxide

CAS: 13463-67-7; EC No: 236-675-5; MDL No: MFCD00011269; RTECS: XR2275000 -22 Mesh Powder; Linear Formula: TiO2; MW: 79.90 Melting Point: 1830-1850°; Boiling Point: 2500-3000° Density (g/mL): 4.26


  • RTECS: XR2275000
  • -22 Mesh Powder

Formula: TiO₂
MW: 79.88 g/mol
Boiling Pt: 2900
Melting Pt: 1855
Density: 4.26 g/mL
MDL Number: MFCD00011269
CAS Number: 13463-67-7
EINECS: 236-675-5
Merck Index: 13,09549

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR