4-Cyano-3-fluorophenylboronic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Cyano-3-fluorobenzeneboronic acid

211350-1G 211350-25G 211350-5G
BT211350-1GEA 85 USD
BT211350-1G BT211350-25G BT211350-5G
4-Cyano-3-fluorophenylboronic acid ≥97%
4-Cyano-3-fluorophenylboronic acid

CAS: 843663-18-3; MDL No: MFCD03411549 Powder; Molecular Formula: C7H5BFNO2; MW: 164.93


  • Powder

Formula: C₇H₅BFNO₂
MW: 164.93 g/mol
MDL Number: MFCD03411549
CAS Number: 843663-18-3

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR