4-Cyano-3-fluorophenylboronic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Cyano-3-fluorobenzeneboronic acid
211350-1G
211350-25G
211350-5G
BT211350-1GEA
85
USD
BT211350-1G
BT211350-25G
BT211350-5G
4-Cyano-3-fluorophenylboronic acid ≥97%
4-Cyano-3-fluorophenylboronic acid
CAS: 843663-18-3; MDL No: MFCD03411549
Powder; Molecular Formula: C7H5BFNO2; MW: 164.93
- Powder
Formula:
C₇H₅BFNO₂ MW: 164.93 g/mol |
MDL Number:
MFCD03411549 CAS Number: 843663-18-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...