Kieselguhr, powder -325 Mesh

Supplier: BeanTown Chemical
Danger

Synonyms: Diatomaceous silica, Diatomaceous earth, Siliceous earth, Infusorial earth, Diatomite, Celite

211300-1KG 211300-5KG
BT211300-1KGEA 42.72 USD
BT211300-1KG BT211300-5KG
Kieselguhr, powder -325 Mesh
Kieselguhr

CAS: 7631-86-9; EC No: 231-545-4; MDL No: MFCD00132803; RTECS: EU8655000 -325 Mesh Powder


  • RTECS: EU8655000

Formula: O₂Si
MDL Number: MFCD00132803
CAS Number: 68855-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR