Disodium tetrasulfide ≥90%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Sodium tetrasulfide, Sodium tetrasulphide, Sodium sulfide, Disodium tetrasulfide, Sodium sulphide

211150-10G 211150-50G
BT211150-10GEA 122 USD
BT211150-10G BT211150-50G
Disodium tetrasulfide ≥90%, Technical Grade
Disodium tetrasulphide

CAS: 12034-39-8; EC No: 215-211-5; MDL No: MFCD00014249 UN No: UN1385; Haz Class: 4.2; Packing Group: II Powder; Molecular Formula: Na2S4 ; MW: 174.24 Melting Point: 275° Hygroscopic, Moisture Sensitive


  • Hygroscopic, Moisture Sensitive
  • Powder

Formula: Na₂S₄
MW: 174.24
Melting Pt: 275 °C
MDL Number: MFCD00014249
CAS Number: 12034-39-8
EINECS: 215-211-5
UN: 1385
ADR: 4.2 II

Order Now

Specification Test Results

Assay 90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR