Bismuth ≥99.99% (trace metals basis), rods Ø 11 mm

Supplier: BeanTown Chemical

211110-100G 211110-25G 211110-500G
BT211110-100GEA 205 USD
BT211110-100G BT211110-25G BT211110-500G
Bismuth ≥99.99% (trace metals basis), rods Ø 11 mm
Bismuth

CAS: 7440-69-9; EC No: 231-177-4; MDL No: MFCD00134033 Solid; Molecular Formula: Bi; MW: 208.98 Melting Point: 271°; Boiling Point: 1560° Density (g/mL): 9.8


  • Solid

Formula: Bi
MW: 208.98 g/mol
Boiling Pt: 1490
Melting Pt: 271 °C
Density: 9.8 g/mL
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR