4-Iodo-2-methylimidazole

Supplier: BeanTown Chemical

Synonyms: 4-Iodo-2-methyl-1H-imidazole

145880-1G 145880-5G
BT145880-1GEA 55 USD
BT145880-1G BT145880-5G
4-Iodo-2-methylimidazole
4-Iodo-2-methylimidazole

CAS: 73746-45-9; MDL No: MFCD15071119 Solid; Molecular Formula: C4H5IN2; MW: 208.00 Melting Point: 133-137°


  • Solid

Formula: C₄H₅IN₂
MW: 208 g/mol
Melting Pt: 133…137 °C
MDL Number: MFCD02684335
CAS Number: 73746-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR