4-Iodo-2-methylimidazole
Supplier: BeanTown Chemical
Synonyms:
4-Iodo-2-methyl-1H-imidazole
145880-1G
145880-5G
BT145880-1GEA
55
USD
BT145880-1G
BT145880-5G
4-Iodo-2-methylimidazole
4-Iodo-2-methylimidazole
CAS: 73746-45-9; MDL No: MFCD15071119
Solid; Molecular Formula: C4H5IN2; MW: 208.00
Melting Point: 133-137°
- Solid
Formula:
C₄H₅IN₂ MW: 208 g/mol Melting Pt: 133…137 °C |
MDL Number:
MFCD02684335 CAS Number: 73746-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...