Barium tetracyanoplatinate(II) tetrahydrate ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

145865-1G 145865-5G
BT145865-1GEA 301 USD
BT145865-1G BT145865-5G
Barium tetracyanoplatinate(II) tetrahydrate ≥99.99% (trace metals basis)
Barium tetracyanoplatinate(II) tetrahydrate

CAS: 13755-32-3; EC No: 209-238-1; MDL No: MFCD00014183 UN No: UN1564; Haz Class: 6.1; Packing Group: II Powder; Linear Formula: BaPt(CN)4·4H2O; MW: 508.56


  • Powder

Formula: BaPt(CN)₄.4H₂O
MDL Number: MFCD00014183
CAS Number: 13755-32-3
EINECS: 209-238-1
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR