DL-Methionine ≥99%
Supplier: BeanTown Chemical
Synonyms:
2-Amino-4-(methylthio)butanoic acid, (±)-2-Amino-4-(methylthio)butyric acid, Methioninum racemicum, Methionine racematic, H-DL-Met-OH, (±)-Methionine, (±)-2-Amino-4-(methylthio)butanoic acid
145805-1KG
145805-250G
145805-5KG
BT145805-1KGEA
76.28
USD
BT145805-1KG
BT145805-250G
BT145805-5KG
DL-Methionine ≥99%
DL-Methionine
CAS: 59-51-8; EC No: 200-432-1; MDL No: MFCD00063097; RTECS: PD0457000
Powder; Molecular Formula: C5H11NO2S; MW: 149.21
Melting Point: 280° (decomposes)
- RTECS: PD0457000
- Powder
Formula:
C₅H₁₁NO₂S Melting Pt: 280 °C (dec.) |
MDL Number:
MFCD00063096 CAS Number: 59-51-8 EINECS: 200-432-1 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...