Iron(II) sulfate heptahydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Iron vitriol, Ferrous sulphate heptahydrate, Ferrous sulfate heptahydrate

145775-10KG 145775-2.5KG 145775-500G
BT145775-10KGEA 290 USD
BT145775-10KG BT145775-2.5KG BT145775-500G
Iron(II) sulfate heptahydrate ≥98%
Iron (II) sulphate heptahydrate

CAS: 7782-63-0; EC No: 231-753-5; MDL No: MFCD00149719; RTECS: NO8510000 Crystalline; Linear Formula: FeSO4·7H2O; MW: 278.02 Density (g/mL): 1.898 Air Sensitive


  • RTECS: NO8510000
  • Air Sensitive
  • Crystalline

Formula: FeSO₄·7H₂O
Density: 1.898 g/mL
MDL Number: MFCD00149719
CAS Number: 7782-63-0
EINECS: 231-753-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR