Dichloromethane ≥99.7% stabilised for spectrophotometry
Supplier: BeanTown Chemical
Synonyms:
Methane dichloride, Methylene chloride, Methylene chloride
145765-1L
145765-4X1L
BT145765-1LEA
68.78
USD
BT145765-1L
BT145765-4L
BT145765-4X1L
Dichloromethane ≥99.7% stabilised for spectrophotometry
Dichloromethane
CAS: 75-09-2; EC No: 200-838-9; MDL No: MFCD00000881; RTECS: PA8050000
UN No: UN1593; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: CH2Cl2; MW: 84.93
Melting Point: -97°; Boiling Point: 39-40°
Density (g/mL): 1.325; Refractive Index: 1.424
- RTECS: PA8050000
- Refractive index: 1.424
- Liquid
Formula:
CH₂Cl₂ Boiling Pt: 39…40 °C Melting Pt: –97 °C Density: 1.325 g/mL |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1,III |
|
Specification Test Results
Assay | 99.7% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...