Copper ≥99.999% (trace metals basis), wire Ø 0.64 mm

Supplier: BeanTown Chemical

145755-10M 145755-2M
BT145755-10MEA 213 USD
BT145755-10M BT145755-2M
Copper ≥99.999% (trace metals basis), wire Ø 0.64 mm
Copper

CAS: 7440-50-8; EC No: 231-159-6; MDL No: MFCD00010965 Solid; Molecular Formula: Cu; MW: 63.55 Melting Point: 1083.4°; Boiling Point: 2567° Density (g/mL): 8.94


  • Solid

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/mL
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR