Pyrite (FeS₂) naturally occurring mineral 7.6x7.6 cm
Supplier: BeanTown Chemical
145705-1PC
145705-5PC
BT145705-1PCEA
179
USD
BT145705-1PC
BT145705-5PC
Pyrite (FeS₂) naturally occurring mineral 7.6x7.6 cm
Pyrite (FeS₂)
CAS: 1309-36-0; EC No: 215-167-7; MDL No: MFCD00064690
Solid; Molecular Formula: FeS2; MW: 119.98
Density (g/mL): 5.02
- Solid
Formula:
FeS₂ Density: 5.02 g/mL |
MDL Number:
MFCD00064690 CAS Number: 1309-36-0 EINECS: 215-167-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...