Pyrite (FeS₂) naturally occurring mineral 7.6x7.6 cm

Supplier: BeanTown Chemical

145705-1PC 145705-5PC
BT145705-1PCEA 179 USD
BT145705-1PC BT145705-5PC
Pyrite (FeS₂) naturally occurring mineral 7.6x7.6 cm
Pyrite (FeS₂)

CAS: 1309-36-0; EC No: 215-167-7; MDL No: MFCD00064690 Solid; Molecular Formula: FeS2; MW: 119.98 Density (g/mL): 5.02


  • Solid

Formula: FeS₂
Density: 5.02 g/mL
MDL Number: MFCD00064690
CAS Number: 1309-36-0
EINECS: 215-167-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR