Tetrahexylammonium hydrogen sulfate ≥98%

Supplier: BeanTown Chemical

145690-1G
BT145690-1GEA 49 USD
BT145690-1G
Tetrahexylammonium hydrogen sulfate ≥98%
Tetrahexylammonium hydrogen sulfate

CAS: 32503-34-7; EC No: 251-069-0; MDL No: MFCD00037675 Crystalline/Powder; Linear Formula: [CH3(CH2)5]4N(HSO4); Molecular Formula: C24H53NO4S ; MW: 451.76 Melting Point: 98-100° Hygroscopic


  • Hygroscopic
  • Crystalline/Powder

Formula: C₂₄H₅₃NO₄S
MW: 451.76
Melting Pt: 98…100 °C
MDL Number: MFCD00037675
CAS Number: 32503-34-7
EINECS: 251-069-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR