Sodium thiophosphate hydrate
Supplier: BeanTown Chemical
145665-25G
145665-5G
BT145665-25GEA
292
USD
BT145665-25G
BT145665-5G
Sodium thiophosphate hydrate
Sodium thiophosphate hydrate
CAS: 10489-48-2; MDL No: MFCD00041513
Solid; Linear Formula: Na3PO3S·xH2O; MW: 180.00 (anhydrous)
Melting Point: 53-58°
- Solid
Formula:
H₂Na₃O₄PS Melting Pt: 53…58 °C |
MDL Number:
MFCD00041513 CAS Number: 10489-48-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...