2,2,2-Tribromoethanol ≥99%

Supplier: BeanTown Chemical

Synonyms: Tribromoethyl alcohol

145585-100G 145585-25G 145585-5G
BT145585-100GEA 261 USD
BT145585-100G BT145585-25G BT145585-5G
2,2,2-Tribromoethanol ≥99%
2,2,2-Tribromoethanol

CAS: 75-80-9; EC No: 200-903-1; MDL No: MFCD00004671; RTECS: KM3675000 Solid; Linear Formula: Br3CCH2OH; Molecular Formula: C2H3Br3O; MW: 282.76 Melting Point: 73-79°; Boiling Point: 92-93°/10 mmHg Light Sensitive


  • RTECS: KM3675000
  • Light Sensitive
  • Solid

Formula: Br₃CCH₂OH
MW: 282.76 g/mol
Boiling Pt: 92…93 °C/10 mmHg
Melting Pt: 81
MDL Number: MFCD00004671
CAS Number: 75-80-9
EINECS: 200-903-1
Merck Index: 12,09743

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR