Zirconium ≥99.5% (trace metals basis excluding Hf), foil, annealed, Thickness 0.25 mm
Supplier: BeanTown Chemical
145490-100X125
145490-125X200
145490-125X500
BT145490-100X1EA
57.28
USD
BT145490-100X1
BT145490-125X2
BT145490-125X5
Zirconium ≥99.5% (trace metals basis excluding Hf), foil, annealed, Thickness 0.25 mm
Zirconium
CAS: 7440-67-7; EC No: 231-176-9; MDL No: MFCD00011303; RTECS: ZH7070000
Solid; Molecular Formula: Zr; MW: 91.22
Melting Point: 1852°; Boiling Point: 4377°
Density (g/mL): 6.5
- RTECS: ZH7070000
- Solid
Formula:
Zr Boiling Pt: 4377 °C Melting Pt: 1852 °C Density: 6.5 g/mL |
MDL Number:
MFCD00011303 CAS Number: 7440-67-7 EINECS: 231-176-9 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...