1-Phenyl-1,3-butandione ≥98%
Supplier: BeanTown Chemical
145435-1KG
BT145435-1KGEA
609
USD
BT145435-1KG
1-Phenyl-1,3-butandione ≥98%
1-Phenyl-1,3-butandione
CAS: 93-91-4; EC No: 202-286-4; MDL No: MFCD00008786; RTECS: EK3540200
Powder; Molecular Formula: C10H10O2; MW: 162.19
Melting Point: 54-56°
Density (g/mL): 1.09
- RTECS: EK3540200
- Powder
Formula:
C₆H₅C(O)CH₂C(O)CH₃ Melting Pt: 54…56 °C Density: 1.09 g/mL |
MDL Number:
MFCD00008786 CAS Number: 93-91-4 EINECS: 202-286-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...