Tetraethylene glycol ≥99%
Supplier: BeanTown Chemical
Synonyms:
2,2'-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanol, Tetraglycol
145420-2.5L
145420-500ML
BT145420-2.5LEA
86
USD
BT145420-2.5L
BT145420-500ML
Tetraethylene glycol ≥99%
Tetraethylene glycol
CAS: 112-60-7; EC No: 203-989-9; MDL No: MFCD00002879; RTECS: XC2100000
Liquid; Linear Formula: HO(CH2CH2O)3CH2CH2OH; Molecular Formula: C8H18O5; MW: 194.23
Melting Point: -5.6°; Boiling Point: 313-314°; Flash point: 193°C (379°F)
Density (g/mL): 1.125 ; Refractive Index: 1.459
- RTECS: XC2100000
- Refractive index: 1.459
- Liquid
Formula:
C₈H₁₈O₅ MW: 194.23 g/mol Boiling Pt: 313…314 °C Melting Pt: –5.6 °C Density: 1.125 g/mL Flash Pt: 193 °C (379 °F) |
MDL Number:
MFCD00002879 CAS Number: 112-60-7 EINECS: 203-989-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...