Sodium sulfite, anhydrous ≥98%

Supplier: BeanTown Chemical

145385-10KG 145385-2.5KG 145385-500G
BT145385-10KGEA 288 USD
BT145385-10KG BT145385-2.5KG BT145385-500G
Sodium sulfite, anhydrous ≥98%
Sodium sulphite

CAS: 7757-83-7; EC No: 231-821-4; MDL No: MFCD00003503; RTECS: WE2150000 Solid; Linear Formula: Na2SO3; MW: 126.04 Melting Point: 500° (decomposes) Density (g/mL): 2.63 Air Sensitive, Moisture Sensitive


  • RTECS: WE2150000
  • Air Sensitive, Moisture Sensitive
  • Solid

Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: >500 °C (dec.)
Density: 2.63 g/mL
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
Merck Index: 13,08757

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR