[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex (1:1)
Supplier: BeanTown Chemical
Synonyms:
Dichloro 1,1'-bis(diphenylphosphino)ferrocene palladium (II) dichloromethane, [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) Dichloride Dichloromethane Adduct (1:1), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with dichloromethane, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane complex
CAS: 95464-05-4; MDL No: MFCD00792899
Powder; Molecular Formula: C34H30Cl4FeP2Pd; MW: 816.64
Melting Point: 275-280°
- Powder
Formula:
C₃₅H₃₀Cl₄FeP₂Pd Melting Pt: 275…280 °C |
MDL Number:
MFCD00792899 CAS Number: 95464-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...