di-Lithium tetraborate ≥99%

Supplier: BeanTown Chemical

Synonyms: Lithium tetraborate, Dilithium tetraborate

145255-1KG 145255-250G 145255-50G
BT145255-1KGEA 536.27 USD
BT145255-1KG BT145255-250G BT145255-50G
di-Lithium tetraborate ≥99%
di-Lithium tetraborate

CAS: 12007-60-2; EC No: 234-514-3; MDL No: MFCD00011083 Powder; Linear Formula: Li2B4O7; MW: 169.12 Melting Point: 760-880°


  • Powder

Formula: Li₂B₄O₇
MW: 169.12 g/mol
Melting Pt: 918
MDL Number: MFCD00011083
CAS Number: 12007-60-2
EINECS: 234-514-3
Merck Index: 14,05524

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR