Sodium hexacyanoferrate(II) decahydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Sodium ferrocyanide decahydrate
145245-250G
145245-50G
BT145245-250GEA
140
USD
BT145245-250G
BT145245-50G
Sodium hexacyanoferrate(II) decahydrate ≥99%
Sodium hexacyanoferrate(II) decahydrate
CAS: 14434-22-1; EC No: 237-081-9; MDL No: MFCD00085360; RTECS: LJ8145000
Powder; Linear Formula: Na4Fe(CN)6·10H2O; Molecular Formula: C6FeN6Na4·10H2O; MW: 484.07
Density (g/mL): 1.458
- RTECS: LJ8145000
- Powder
Formula:
C₆H₂₀FeN₆Na₄O₁₀ Density: 1.458 g/mL |
MDL Number:
MFCD00085360 CAS Number: 14434-22-1 EINECS: 237-081-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...