Maltitol ≥97%
Supplier: BeanTown Chemical
Synonyms:
(2S,3R,4R,5R)-4-(((2R,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexane-1,2,3,5,6-pentaol
145215-1KG
145215-250G
145215-50G
BT145215-1KGEA
262
USD
BT145215-1KG
BT145215-250G
BT145215-50G
Maltitol ≥97%
Maltitol
CAS: 585-88-6; EC No: 209-567-0; MDL No: MFCD00006600; RTECS: LZ4394000
Crystalline/Powder; Molecular Formula: C12H24O11; MW: 344.31
Melting Point: 149-152°
- RTECS: LZ4394000
- Crystalline/Powder
Formula:
C₁₂H₂₄O₁₁ Melting Pt: 149…152 °C |
MDL Number:
MFCD00006600 CAS Number: 585-88-6 EINECS: 209-567-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...