N,N'-Di-n-propylethylenediamine ≥97%
Supplier: BeanTown Chemical
Synonyms:
N1,N1-Dipropylethane-1,2-diamine, N,N'-Di(n-propyl)ethylenediamine
145205-25G
145205-5G
BT145205-25GEA
234
USD
BT145205-25G
BT145205-5G
N,N'-Di-n-propylethylenediamine ≥97%
N,N'-Di-n-propylethylenediamine
CAS: 14165-22-1; MDL No: MFCD00015199
UN No: UN2735; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C8H20N2; MW: 144.26
Boiling Point: 186-189°; Flash point: 69°C (156°F)
Density (g/mL): 0.802; Refractive Index: 1.440
Air Sensitive
- Refractive index: 1.440
- Air Sensitive
- Liquid
Formula:
C₈H₂₀N₂ MW: 144.262 g/mol Boiling Pt: 186…189 °C Density: 0.802 g/mL Flash Pt: 69 °C (156 °F) |
MDL Number:
MFCD00015199 CAS Number: 14165-22-1 ADR: 8 III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...