Methyltin trichloride ≥97%

Supplier: BeanTown Chemical

Synonyms: Trichloromethylstannane

145190-1G 145190-25G 145190-5G
BT145190-1GEA 48 USD
BT145190-1G BT145190-25G BT145190-5G
Methyltin trichloride ≥97%
Methyltin trichloride

CAS: 993-16-8; EC No: 213-608-8; MDL No: MFCD00000514 UN No: UN2923; Haz Class: 8 (6.1); Packing Group: II Crystalline; Linear Formula: CH3SnCl3; MW: 240.11 Melting Point: 48-51°; Boiling Point: 171°; Flash point: 41°C (106°F) Moisture Sensitive


  • Moisture Sensitive
  • Crystalline

Formula: CH₃Cl₃Sn
Boiling Pt: 171 °C
Melting Pt: 48…51 °C
Flash Pt: 41 °C (106 °F)
MDL Number: MFCD00000514
CAS Number: 993-16-8
EINECS: 213-608-8
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR