3-Amino-1H-1,2,4-triazole-5-thiol ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-Amino-5-mercapto-1,2,4-triazole, 3-Amino-1,2,4-triazole-5-thiol, 3-Amino-4H-1,2,4-triazole-5-thiol, 3-Amino-5-mercapto-1,2,4-triazole 5-Amino-4H-1,2,4-triazole-3-thiol

145165-100G 145165-25G 145165-5G
BT145165-100GEA 366 USD
BT145165-100G BT145165-25G BT145165-5G
3-Amino-1H-1,2,4-triazole-5-thiol ≥98%
3-Amino-1H-1,2,4-triazole-5-thiol

CAS: 16691-43-3; EC No: 240-735-6; MDL No: MFCD00005231; RTECS: XZ5357500 Powder; Molecular Formula: C2H4N4; MW: 116.14 Melting Point: 300°


  • RTECS: XZ5357500
  • Powder

Formula: C₂H₄N₄S
Melting Pt: >300 °C
MDL Number: MFCD00005231
CAS Number: 16691-43-3
EINECS: 240-735-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR