2-Chloro-5-(trifluoromethyl)-4-iodopyridine ≥95%

Supplier: BeanTown Chemical

Synonyms: 2-Chloro-4-iodo-5-(trifluoromethyl)pyridine

145145-1G 145145-250MG 145145-5G
BT145145-1GEA 118 USD
BT145145-1G BT145145-250MG BT145145-5G
2-Chloro-5-(trifluoromethyl)-4-iodopyridine ≥95%
2-Chloro-5-(trifluoromethyl)-4-iodopyridine

CAS: 505084-55-9 Solid; Molecular Formula: C6H2ClF3IN; MW: 307.44 Light Sensitive


  • Light Sensitive
  • Solid

Formula: C₆H₂ClF₃IN
MW: 307.44 g/mol
MDL Number: MFCD01862099
CAS Number: 505084-55-9

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR