2,6-Dihydroxynaphthalene ≥98%
Supplier: BeanTown Chemical
Synonyms:
Naphthalene-2,6-diol
145115-25G
145115-5G
BT145115-25GEA
246
USD
BT145115-25G
BT145115-5G
2,6-Dihydroxynaphthalene ≥98%
2,6-Dihydroxynaphthalene
CAS: 581-43-1; EC No: 209-465-6; MDL No: MFCD00004082
Crystalline; Molecular Formula: C10H8O2; MW: 160.17
Melting Point: 223-225°
- Crystalline
Formula:
C₁₀H₈O₂ Melting Pt: 223…225 °C |
MDL Number:
MFCD00004082 CAS Number: 581-43-1 EINECS: 209-465-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...