Bromophenol blue ACS
Supplier: BeanTown Chemical
Synonyms:
Bromphenol Blue Sultone Form, 3,3',5,5'-Tetrabromophenolsulfonphthalein, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein, Tetrabromophenol-sulfonephthalein
145060-1G
145060-25G
145060-5G
BT145060-1GEA
11.55
USD
BT145060-1G
BT145060-25G
BT145060-5G
Bromophenol blue ACS
Bromophenol blue
CAS: 115-39-9; EC No: 204-086-2; MDL No: MFCD00005875; RTECS: SJ7453000
Crystalline; Molecular Formula: C19H10Br4O5S; MW: 669.96
Melting Point: 273° (decomposes)
- RTECS: SJ7453000
- Crystalline
Formula:
C₁₉H₁₀Br₄O₅S Melting Pt: 273 °C (dec.) |
MDL Number:
MFCD00005875 CAS Number: 115-39-9 EINECS: 204-086-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...