Aluminium ≥99.9% (trace metals basis), granules 8-12 mm
Supplier: BeanTown Chemical
145050-1KG
145050-250G
145050-5KG
BT145050-1KGEA
74.51
USD
BT145050-1KG
BT145050-250G
BT145050-5KG
Aluminium ≥99.9% (trace metals basis), granules 8-12 mm
Aluminium
CAS: 7429-90-5; EC No: 231-072-3; MDL No: MFCD00134029
Solid; Molecular Formula: Al; MW: 26.98
Melting Point: 660.4°; Boiling Point: 2460°
Density (g/mL): 2.7
- Solid
Formula:
Al Boiling Pt: 2460 °C Melting Pt: 660.4 °C Density: 2.7 g/mL |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...