(R)-(-)-Hydratropic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: (R)-(-)-2-Phenylpropanoic acid, (R)-(-)-2-Phenylpropionic acid, (R)-2-Phenylpropanoic acid

144965-1G 144965-250MG 144965-5G
BT144965-1GEA 176 USD
BT144965-1G BT144965-250MG BT144965-5G
(R)-(-)-Hydratropic acid ≥97%
(R)-(-)-Hydratropic acid

CAS: 7782-26-5; MDL No: MFCD00063140 Crystalline; Molecular Formula: C9H10O2 ; MW: 150.17 Melting Point: 29-30°; Boiling Point: 115°/1 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.1; Optical Rotation: [α]20/D −72°, c = 1.6 in chloroform


  • Optical rotation: [α]20/D −72°, c = 1.6 in chloroform
  • Crystalline

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 257
Melting Pt: 29…30 °C
Density: 1.1 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00063140
CAS Number: 7782-26-5

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR