1,6-Heptadiyne ≥97%

Supplier: BeanTown Chemical

144950-100G 144950-25G 144950-5G
BT144950-100GEA 1440 USD
BT144950-100G BT144950-25G BT144950-5G
1,6-Heptadiyne ≥97%
1,6-Heptadiyne

CAS: 2396-63-6; EC No: 219-253-5; MDL No: MFCD00014925; RTECS: MI5600000 UN No: UN3295; Haz Class: 3; Packing Group: II Liquid; Linear Formula: HC≡C(CH2)3C≡CH; Molecular Formula: C7H8; MW: 92.14 Boiling Point: 112°; Flash point: 9°C (48°F) Density (g/mL): 0.805; Refractive Index: 1.441


  • RTECS: MI5600000
  • Refractive index: 1.441
  • Liquid

Formula: C₇H₈
MW: 92.14 g/mol
Boiling Pt: 108
Melting Pt: -85
Density: 0.805 g/mL
Flash Pt: 9 °C (48 °F)
MDL Number: MFCD00014925
CAS Number: 2396-63-6
EINECS: 219-253-5
UN: 3295
ADR: 3 II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR