n-Tetradecane ≥99%
Supplier: BeanTown Chemical
CAS: 629-59-4; EC No: 211-096-0; MDL No: MFCD00008986; RTECS: XB8000000
Liquid; Linear Formula: CH3(CH2)12CH3; Molecular Formula: C14H30; MW: 198.39
Melting Point: 6°; Boiling Point: 252-254°; Flash point: 99°C (211°F)
Density (g/mL): 0.762; Refractive Index: 1.429
- RTECS: XB8000000
- Refractive index: 1.429
- Liquid
Formula:
H₃C(CH₂)₁₂CH₃ Boiling Pt: 252…254 °C Melting Pt: 6 °C Density: 0.762 g/mL Flash Pt: 99 °C (211 °F) |
MDL Number:
MFCD00008986 CAS Number: 629-59-4 EINECS: 211-096-0 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...