n-Tetradecane ≥99%

Supplier: BeanTown Chemical

144865-100G 144865-25G 144865-500G
BT144865-100GEA 53.33 USD
BT144865-100G BT144865-25G BT144865-500G
n-Tetradecane ≥99%
n-Tetradecane

CAS: 629-59-4; EC No: 211-096-0; MDL No: MFCD00008986; RTECS: XB8000000 Liquid; Linear Formula: CH3(CH2)12CH3; Molecular Formula: C14H30; MW: 198.39 Melting Point: 6°; Boiling Point: 252-254°; Flash point: 99°C (211°F) Density (g/mL): 0.762; Refractive Index: 1.429


  • RTECS: XB8000000
  • Refractive index: 1.429
  • Liquid

Formula: H₃C(CH₂)₁₂CH₃
Boiling Pt: 252…254 °C
Melting Pt: 6 °C
Density: 0.762 g/mL
Flash Pt: 99 °C (211 °F)
MDL Number: MFCD00008986
CAS Number: 629-59-4
EINECS: 211-096-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03064a29015d1e810f1e4350a7a6cd2dfe4713fd&vid=f4838a5686fe033622495a3bf280963555654d9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR