5-Bromoanthranilic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Amino-5-bromobenzoic acid

144700-100G 144700-25G 144700-5G
BT144700-100GEA 616 USD
BT144700-100G BT144700-25G BT144700-5G
5-Bromoanthranilic acid ≥98%
5-Bromoanthranilic acid

CAS: 5794-88-7; EC No: 227-338-3; MDL No: MFCD00007823; RTECS: CB2557670 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C7H5BrNO2; MW: 216.03 Melting Point: 213-215° Light Sensitive


  • RTECS: CB2557670
  • Light Sensitive
  • Powder

Formula: C₇H₆BrNO₂
Melting Pt: 213…215 °C
MDL Number: MFCD00007823
CAS Number: 5794-88-7
EINECS: 227-338-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR