5-Bromoanthranilic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Amino-5-bromobenzoic acid
144700-100G
144700-25G
144700-5G
BT144700-100GEA
616
USD
BT144700-100G
BT144700-25G
BT144700-5G
5-Bromoanthranilic acid ≥98%
5-Bromoanthranilic acid
CAS: 5794-88-7; EC No: 227-338-3; MDL No: MFCD00007823; RTECS: CB2557670
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C7H5BrNO2; MW: 216.03
Melting Point: 213-215°
Light Sensitive
- RTECS: CB2557670
- Light Sensitive
- Powder
Formula:
C₇H₆BrNO₂ Melting Pt: 213…215 °C |
MDL Number:
MFCD00007823 CAS Number: 5794-88-7 EINECS: 227-338-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...