Acetic acid-[D₄] ≥99.5% (Isotopic)

Supplier: BeanTown Chemical

Synonyms: Ethanoic acid-D4, Acetic-D3 acid-D, Acetic-d3 acid-d, Acetic acid-[²H₄], Tetradeuteroacetic acid, Acetic acid-d₄

144645-10G 144645-5G
BT144645-10GEA 154.11 USD
BT144645-10G BT144645-5G
Acetic acid-[D₄] ≥99.5% (Isotopic)
Acetic acid-[D4]

CAS: 1186-52-3; EC No: 214-693-4; MDL No: MFCD00051051 UN No: UN2789; Haz Class: 8 (3); Packing Group: II Liquid; Linear Formula: CD3CO2D; MW: 64.08 Melting Point: 15-16°; Boiling Point: 115.5°; Flash point: 40°C (104°F) Density (g/mL): 1.119; Refractive Index: 1.368 Moisture Sensitive


  • Refractive index: 1.368
  • Moisture Sensitive
  • Liquid

Formula: D₃CCOOD
MW: 64.02 g/mol
Boiling Pt: 115.5
Melting Pt: 16
Density: 1.119 g/mL
Flash Pt: 40 °C (104 °F)
MDL Number: MFCD00051051
CAS Number: 1186-52-3
EINECS: 214-693-4
UN: 2789
ADR: 8 II

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR