Potassium permanganate ≥98%
Supplier: BeanTown Chemical
CAS: 7722-64-7; EC No: 231-760-3; MDL No: MFCD00011364; RTECS: SD6475000
UN No: UN1490; Haz Class: 5.1; Packing Group: II
Crystalline; Molecular Formula: KMnO4; MW: 158.03
Melting Point: 240°
Density (g/mL): 2.71
Light Sensitive
- RTECS: SD6475000
- Light Sensitive
- Crystalline
Formula:
KMnO₄ Melting Pt: 240 °C Density: 2.71 g/mL |
MDL Number:
MFCD00011364 CAS Number: 7722-64-7 EINECS: 231-760-3 UN: 1490 ADR: 5.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...