2-Indolinone ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Oxindole, Oxindole, Indolin-2-one, 2,3-Dihydro-2-indolone

144520-100G 144520-25G 144520-5G
BT144520-100GEA 418 USD
BT144520-100G BT144520-25G BT144520-5G
2-Indolinone ≥97%
2-Indolinone

CAS: 59-48-3; EC No: 200-429-5; MDL No: MFCD00005711; RTECS: NM2080500 Crystalline; Molecular Formula: C8H7NO; MW: 133.15 Melting Point: 123-128°; Boiling Point: 227°/73 mmHg


  • RTECS: NM2080500
  • Crystalline

Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 313
Melting Pt: 123…128 °C
MDL Number: MFCD00005711
CAS Number: 59-48-3
EINECS: 200-429-5

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR