2-Indolinone ≥97%
Supplier: BeanTown Chemical
Synonyms:
2-Oxindole, Oxindole, Indolin-2-one, 2,3-Dihydro-2-indolone
144520-100G
144520-25G
144520-5G
BT144520-100GEA
418
USD
BT144520-100G
BT144520-25G
BT144520-5G
2-Indolinone ≥97%
2-Indolinone
CAS: 59-48-3; EC No: 200-429-5; MDL No: MFCD00005711; RTECS: NM2080500
Crystalline; Molecular Formula: C8H7NO; MW: 133.15
Melting Point: 123-128°; Boiling Point: 227°/73 mmHg
- RTECS: NM2080500
- Crystalline
Formula:
C₈H₇NO MW: 133.15 g/mol Boiling Pt: 313 Melting Pt: 123…128 °C |
MDL Number:
MFCD00005711 CAS Number: 59-48-3 EINECS: 200-429-5 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...