Aluminium oxide ≥99.995% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

144515-100G 144515-25G 144515-500G
BT144515-100GEA 412 USD
BT144515-100G BT144515-25G BT144515-500G
Aluminium oxide ≥99.995% (trace metals basis)
Aluminium oxide

CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424 Powder; Molecular Formula: Al2O3; MW: 101.96 Melting Point: 2040°


  • Powder

Formula: Al₂O₃
Melting Pt: 2040 °C
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR