Digallium triselenide ≥99.99%
Supplier: BeanTown Chemical
Synonyms:
Gallium (III) selenide
144505-1G
144505-5G
BT144505-1GEA
35
USD
BT144505-1G
BT144505-5G
Digallium triselenide ≥99.99%
Digallium triselenide
CAS: 12024-24-7; MDL No: MFCD00016103
UN No: UN3077; Haz Class: 9; Packing Group: III
3-12mm Sintered Lump; Molecular Formula: Ga2Se3; MW: 376.33
- 3-12mm Sintered Lump
Formula:
Ga₂Se₃ |
MDL Number:
MFCD00016103 CAS Number: 12024-24-7 ADR: 9,III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...