Trimethyl orthobutyrate ≥97%

Supplier: BeanTown Chemical

Synonyms: Trimethylorthobutyrate, 1,1,1-Trimethoxybutane, Orthobutyric Acid Trimethyl Ester

144485-25G 144485-5G
BT144485-25GEA 148 USD
BT144485-25G BT144485-5G
Trimethyl orthobutyrate ≥97%
Trimethyl orthobutyrate

CAS: 43083-12-1; EC No: 256-079-9; MDL No: MFCD00008480 UN No: UN3272; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3CH2CH2C(OCH3)3; Molecular Formula: C7H16O3; MW: 148.20 Boiling Point: 145-157°; Flash point: 50°C (122°F) Density (g/mL): 0.926; Refractive Index: 1.404 Moisture Sensitive


  • Refractive index: 1.404
  • Moisture Sensitive
  • Liquid

Formula: C₇H₁₆O₃
MW: 148.20 g/mol
Boiling Pt: 145…157 °C
Density: 0.926 g/mL
Flash Pt: 50 °C (122 °F)
MDL Number: MFCD00008480
CAS Number: 43083-12-1
EINECS: 256-079-9
ADR: 3 III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR